Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019397.3 | NP_062270.1 | 2 - 3 | 367 | 61 | |
GenBank mRNA | AJ245672.1 | - | 2 - 3 | 369 | 61 | |
AK053738.1 | - | 2 - 3 | 364 | 61 | ||
AK078534.1 | - | 2 - 3 | 345 | 61 | ||
BC117702.2 | - | 2 - 3 | 353 | 40062530 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |