Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019680.2 | NP_062654.2 | 6 - 7 | 882 | 77 | |
GenBank mRNA | AF016714.1 | - | 5 - 6 | 823 | 77 | |
AK155203.1 | - | 6 - 7 | 1123 | 77 | ||
AK155709.1 | - | 6 - 7 | 883 | 77 | ||
BC109160.1 | - | 5 - 6 | 814 | 40047677 | 77 | |
BC109171.1 | - | 5 - 6 | 814 | 40048316 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |