Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Habp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019986.3 | NP_064370.2 | 4 - 5 | 864 | 89 | |
GenBank mRNA | AF227684.1 | - | 4 - 5 | 807 | 89 | |
AK016800.1 | - | 4 - 5 | 907 | 89 | ||
AK144206.1 | - | 4 - 5 | 848 | 89 | ||
AK146525.1 | - | 4 - 5 | 837 | 89 | ||
BC082806.1 | - | 4 - 5 | 866 | 89 | ||
BC119460.2 | - | 3 - 4 | 361 | 89 | ||
BC148304.2 | - | 3 - 4 | 368 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |