Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021292.2 | NP_067267.2 | 12 - 13 | 1860 | 128 | |
GenBank mRNA | AJ250841.1 | - | 12 - 13 | 1840 | 128 | |
AK039759.1 | - | 12 - 13 | 1830 | 128 | ||
AK148262.1 | - | 12 - 13 | 1833 | 128 | ||
BC021392.1 | - | 12 - 13 | 1849 | 5069005 | 128 | |
BC021464.1 | - | 12 - 13 | 1849 | 5069004 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |