Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021398.3 | NP_067373.2 | 9 - 10 | 1352 | 106 | |
GenBank mRNA | AF188622.1 | - | 9 - 10 | 1346 | 106 | |
BC089603.1 | - | 8 - 9 | 1052 | 6758493 | 106 | |
BU516072.1 | - | 3 - 4 | 388 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |