Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eed | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021876.3 | NP_068676.1 | 5 - 6 | 1006 | 108 | |
GenBank mRNA | AK077664.1 | - | 5 - 6 | 591 | 108 | |
AK087924.1 | - | 5 - 6 | 568 | 108 | ||
AK131976.1 | - | 5 - 6 | 573 | 108 | ||
AK133890.1 | - | 5 - 6 | 567 | 108 | ||
BC012966.1 | - | 5 - 6 | 1005 | 3991086 | 108 | |
U78103.1 | - | 5 - 6 | 1008 | 108 | ||
U97675.1 | - | 5 - 6 | 1013 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |