Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022007.1 | NP_071290.1 | 2 - 3 | 133 | 69 | |
GenBank mRNA | AI892161.1 | - | 2 - 3 | 133 | 69 | |
AK004113.1 | - | 2 - 3 | 142 | 69 | ||
BC048737.1 | - | 2 - 3 | 146 | 69 | ||
BC061101.1 | - | 2 - 3 | 146 | 6531288 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |