Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eral1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022313.2 | NP_071708.2 | 4 - 5 | 560 | 84 | |
GenBank mRNA | AB049389.1 | - | 4 - 5 | 536 | 84 | |
AF190730.1 | - | 4 - 5 | 528 | 84 | ||
AF190731.1 | - | 4 - 5 | 545 | 84 | ||
AK012155.1 | - | 4 - 5 | 550 | 84 | ||
BC037603.1 | - | 4 - 5 | 545 | 84 | ||
BC068271.1 | - | 4 - 5 | 555 | 30008388 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |