Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100030082.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030082.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030082.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030082.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030082.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030091.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030091.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030091.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030091.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030091.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030083.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030083.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030083.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030083.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030083.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030085.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030085.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030085.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030085.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030085.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030089.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030089.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030089.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030089.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030089.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030094.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030094.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030094.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030094.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030094.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030081.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030081.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030081.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030081.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030081.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030102.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030102.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030102.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030099.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030099.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030099.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030079.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030079.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030079.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030079.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030079.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030086.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030086.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030086.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030086.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030086.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030090.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030090.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030090.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030090.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030090.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030096.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030096.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030096.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030096.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030096.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030103.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030103.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030103.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030095.mm.1 |
NM_001163290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030095.mm.1 |
NM_001163290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030095.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030095.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030095.mm.1 |
TC0100003662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030109.mm.1 |
NM_023341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030109.mm.1 |
NM_023341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030109.mm.1 |
TC0100003662.mm.1 |
MTA TC |