Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023395.2 | NP_075884.1 | 3 - 4 | 569 | 95 | |
GenBank mRNA | AK009971.1 | - | 3 - 4 | 572 | 95 | |
AK018575.1 | - | 3 - 4 | 430 | 95 | ||
AK132541.1 | - | 3 - 4 | 570 | 95 | ||
AK142707.1 | - | 3 - 4 | 438 | 95 | ||
AK161606.1 | - | 3 - 4 | 435 | 95 | ||
BC038122.1 | - | 3 - 4 | 411 | 95 | ||
BC132073.1 | - | 3 - 4 | 498 | 40130350 | 95 | |
BC132469.1 | - | 3 - 4 | 498 | 40130746 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |