Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023596.3 | NP_076085.1 | 2 - 3 | 342 | 99 | |
GenBank mRNA | AF326986.1 | - | 2 - 3 | 353 | 99 | |
AK034133.1 | - | 2 - 3 | 338 | 99 | ||
AK157868.1 | - | 2 - 3 | 343 | 99 | ||
BC137864.1 | - | 2 - 3 | 325 | 8860883 | 99 | |
BC144983.1 | - | 2 - 3 | 325 | 99 | ||
CV557129.1 | - | 1 - 2 | 212 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |