Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023622.4 | NP_076111.2 | 5 - 6 | 699 | 88 | |
GenBank mRNA | AF237669.1 | - | 5 - 6 | 658 | 88 | |
AF239661.1 | - | 5 - 6 | 700 | 88 | ||
AK004876.1 | - | 4 - 5 | 432 | 88 | ||
AK053550.1 | - | 5 - 6 | 686 | 88 | ||
AK131140.1 | - | 107 | 88 | |||
AK134654.1 | - | 681 | 88 | |||
AK165023.1 | - | 5 - 6 | 687 | 88 | ||
AK165746.1 | - | 5 - 6 | 692 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |