Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spock3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252620.1 | NP_001239549.1 | 6 - 7 | 556 | 70 | |
NM_001252621.1 | NP_001239550.1 | 5 - 6 | 547 | 70 | ||
NM_023689.3 | NP_076178.1 | 6 - 7 | 670 | 70 | ||
GenBank mRNA | AJ278998.1 | - | 6 - 7 | 564 | 70 | |
AK013644.1 | - | 6 - 7 | 599 | 70 | ||
AK049843.1 | - | 5 - 6 | 548 | 70 | ||
AK220280.1 | - | 6 - 7 | 654 | 70 | ||
BC017601.1 | - | 6 - 7 | 569 | 4502169 | 70 | |
BC053334.1 | - | 6 - 7 | 541 | 6492113 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |