Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P161342 |
AK211025.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P161342 |
AK217592.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P161342 |
BC006634.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P161342 |
AK164003.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P161342 |
AK168642.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P161342 |
AK211025.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P161342 |
AK217592.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P161342 |
BC006634.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P161342 |
AK164003.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P161342 |
AK168642.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
756809 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930840 |
AB024303.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930840 |
AK010313.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930840 |
AK011783.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930840 |
AK087556.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930840 |
AK090012.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930840 |
AK159672.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930840 |
BC068164.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930840 |
NM_019748.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
687452 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
223443 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
18440 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
952144 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
146648 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P143387 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P161342 |
AK211025.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P161342 |
AK217592.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P161342 |
BC006634.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P161342 |
AK164003.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P161342 |
AK168642.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P161342 |
AK211025.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P161342 |
AK217592.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P161342 |
BC006634.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P161342 |
AK164003.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P161342 |
AK168642.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P161342 |
AK211025.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P161342 |
AK217592.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P161342 |
BC006634.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P161342 |
AK164003.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P161342 |
AK168642.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P161342 |
AK211025.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P161342 |
AK217592.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P161342 |
BC006634.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P161342 |
AK164003.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P161342 |
AK168642.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352453 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352456 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352466 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352457 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352450 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352447 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352448 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352475 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352460 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352462 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352474 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352472 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352458 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352463 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352464 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352454 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352444 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352468 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20352470 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P143387 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04149698 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P161342 |
AK211025.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161342 |
AK217592.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161342 |
BC006634.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161342 |
AK164003.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161342 |
AK168642.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161342 |
AK211025.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161342 |
AK217592.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161342 |
BC006634.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161342 |
AK164003.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P161342 |
AK168642.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P138262 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P143387 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00809679 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00809683 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00809681 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352444 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352355 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352371 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352456 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352372 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352369 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352416 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352435 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352460 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352381 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352379 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352417 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352520 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352375 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352365 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352356 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352385 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352447 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352392 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352401 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352453 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352399 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352412 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352359 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352514 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352448 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352429 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352414 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352383 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352525 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20352457 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00809679 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00462535 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352448 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352442 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352409 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352462 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352406 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00346529 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00462522 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352401 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00204408 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352416 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352426 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00056337 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089960 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00056347 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00462537 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352422 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00462521 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352428 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00346526 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00754881 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00462534 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00346533 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00346534 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00346517 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00204386 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352403 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352460 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00809695 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089958 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00809670 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352435 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352431 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352402 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00809686 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00062391 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00204390 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352419 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352433 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352450 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352437 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00809681 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20352444 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00232598 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00346520 |
- |
- |