Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023773.2 | NP_076262.2 | 7 - 8 | 1886 | 75 | |
GenBank mRNA | AB041598.2 | - | 7 - 8 | 1887 | 75 | |
AK016067.1 | - | 2 - 3 | 131 | 75 | ||
AK158773.1 | - | 7 - 8 | 1864 | 75 | ||
AK159495.1 | - | 7 - 8 | 1864 | 75 | ||
BC079558.1 | - | 7 - 8 | 1819 | 75 | ||
BC145164.1 | - | 7 - 8 | 1811 | 75 | ||
BC145700.1 | - | 7 - 8 | 1811 | 40131003 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |