Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023831.3 | NP_076320.2 | 5 - 6 | 550 | 66 | |
GenBank mRNA | AJ249981.1 | - | 5 - 6 | 594 | 66 | |
AK005360.1 | - | 5 - 6 | 558 | 66 | ||
AK032880.1 | - | 5 - 6 | 551 | 66 | ||
BC010326.1 | - | 4 - 5 | 349 | 3599905 | 66 | |
BC080764.1 | - | 5 - 6 | 492 | 6488976 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |