Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026165.3 | NP_080441.1 | 6 - 7 | 708 | 115 | |
GenBank mRNA | AK004625.1 | - | 6 - 7 | 712 | 115 | |
AK006400.1 | - | 5 - 6 | 344 | 115 | ||
AK008131.1 | - | 6 - 7 | 712 | 115 | ||
AK030346.1 | - | 6 - 7 | 709 | 115 | ||
AK031117.1 | - | 6 - 7 | 709 | 115 | ||
AK078383.1 | - | 718 | 115 | |||
AK147936.1 | - | 6 - 7 | 723 | 115 | ||
AK160834.1 | - | 6 - 7 | 709 | 115 | ||
AK161372.1 | - | 431 | 115 | |||
BC020087.1 | - | 6 - 7 | 692 | 3990909 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |