Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026395.1 | NP_080671.1 | 1 - 2 | 98 | 95 | |
GenBank mRNA | AK002550.1 | - | 1 - 2 | 98 | 95 | |
AK003682.1 | - | 1 - 2 | 120 | 95 | ||
AK013262.1 | - | 1 - 2 | 155 | 95 | ||
AK078396.1 | - | 1 - 2 | 161 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons |