Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026402.3 | NP_080678.1 | 1 - 2 | 348 | 90 | |
GenBank mRNA | AB079386.1 | - | 1 - 2 | 77 | 90 | |
AK005266.1 | - | 1 - 2 | 355 | 90 | ||
AK011409.1 | - | 1 - 2 | 349 | 90 | ||
AK078877.1 | - | 1 - 2 | 341 | 90 | ||
AK150914.1 | - | 1 - 2 | 349 | 90 | ||
AK159231.1 | - | 1 - 2 | 349 | 90 | ||
BC010809.1 | - | 1 - 2 | 346 | 4241199 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |