Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026402.3 | NP_080678.1 | 4 - 5 | 506 | 125 | |
GenBank mRNA | AB079386.1 | - | 4 - 5 | 235 | 125 | |
AK005266.1 | - | 4 - 5 | 513 | 125 | ||
AK011409.1 | - | 4 - 5 | 507 | 125 | ||
AK078877.1 | - | 4 - 5 | 499 | 125 | ||
AK150914.1 | - | 4 - 5 | 507 | 125 | ||
AK159231.1 | - | 4 - 5 | 507 | 125 | ||
BC010809.1 | - | 4 - 5 | 504 | 4241199 | 125 | |
BC030053.1 | - | 2 - 3 | 1228 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |