Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026402.3 | NP_080678.1 | 10 - 11 | 1067 | 78 | |
GenBank mRNA | AB079386.1 | - | 10 - 11 | 796 | 78 | |
AK005266.1 | - | 10 - 11 | 1074 | 78 | ||
AK011409.1 | - | 10 - 11 | 1068 | 78 | ||
AK078877.1 | - | 10 - 11 | 1060 | 78 | ||
AK150914.1 | - | 10 - 11 | 1068 | 78 | ||
AK159231.1 | - | 10 - 11 | 1068 | 78 | ||
AK209780.1 | - | 4 - 5 | 345 | 78 | ||
AK213960.1 | - | 4 - 5 | 339 | 78 | ||
BC010809.1 | - | 10 - 11 | 1065 | 4241199 | 78 | |
BC030053.1 | - | 8 - 9 | 1789 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |