Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026403.3 | NP_080679.3 | 7 - 8 | 1493 | 130 | |
GenBank mRNA | AK011570.1 | - | 7 - 8 | 1483 | 130 | |
AK015585.1 | - | 7 - 8 | 1243 | 130 | ||
AK032873.1 | - | 7 - 8 | 1477 | 130 | ||
AK037831.1 | - | 7 - 8 | 1481 | 130 | ||
AK050912.1 | - | 7 - 8 | 1484 | 130 | ||
AK083861.1 | - | 7 - 8 | 1362 | 130 | ||
AK139689.1 | - | 7 - 8 | 1444 | 130 | ||
AK146020.1 | - | 7 - 8 | 1448 | 130 | ||
AK146687.1 | - | 7 - 8 | 1465 | 130 | ||
AK152732.1 | - | 7 - 8 | 1467 | 130 | ||
AK154387.1 | - | 7 - 8 | 1485 | 130 | ||
AK159798.1 | - | 7 - 8 | 1467 | 130 | ||
BC018523.1 | - | 7 - 8 | 1473 | 3499986 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |