Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026438.4 | NP_080714.2 | 9 - 10 | 907 | 60 | |
GenBank mRNA | AK008575.1 | - | 9 - 10 | 908 | 60 | |
AK020291.1 | - | 9 - 10 | 923 | 60 | ||
AK088397.1 | - | 9 - 10 | 903 | 60 | ||
AK145640.1 | - | 9 - 10 | 957 | 60 | ||
AK146300.1 | - | 9 - 10 | 870 | 60 | ||
AK151512.1 | - | 9 - 10 | 898 | 60 | ||
AK216005.1 | - | 3 - 4 | 261 | 60 | ||
BC010468.1 | - | 9 - 10 | 841 | 3585780 | 60 | |
CT010203.1 | - | 9 - 10 | 800 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |