Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galntl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026449.3 | NP_080725.2 | 2 - 3 | 392 | 80 | |
GenBank mRNA | AK005605.1 | - | 2 - 3 | 413 | 80 | |
AK006193.1 | - | 2 - 3 | 371 | 80 | ||
AK016415.1 | - | 2 - 3 | 393 | 80 | ||
BC049554.1 | - | 2 - 3 | 374 | 80 | ||
BC100447.1 | - | 2 - 3 | 245 | 80 | ||
BC145757.1 | - | 2 - 3 | 252 | 40131060 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |