Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galntl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026449.3 | NP_080725.2 | 7 - 8 | 1167 | 77 | |
GenBank mRNA | AK005605.1 | - | 7 - 8 | 1188 | 77 | |
AK006193.1 | - | 7 - 8 | 1146 | 77 | ||
AK016415.1 | - | 7 - 8 | 1168 | 77 | ||
BC049554.1 | - | 7 - 8 | 1149 | 77 | ||
BC100447.1 | - | 7 - 8 | 1020 | 77 | ||
BC145757.1 | - | 7 - 8 | 1027 | 40131060 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |