Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026486.3 | NP_080762.1 | 2 - 3 | 323 | 82 | |
GenBank mRNA | AK012720.1 | - | 2 - 3 | 296 | 82 | |
AK014476.1 | - | 2 - 3 | 289 | 82 | ||
AK031829.1 | - | 2 - 3 | 324 | 82 | ||
AK032378.1 | - | 1 - 2 | 459 | 82 | ||
AK087856.1 | - | 2 - 3 | 281 | 82 | ||
AK162182.1 | - | 81 | 82 | |||
AK165102.1 | - | 2 - 3 | 289 | 82 | ||
DQ278869.1 | - | 2 - 3 | 321 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |