Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026486.3 | NP_080762.1 | 15 - 16 | 1885 | 61 | |
GenBank mRNA | AK008223.1 | - | 4 - 5 | 61 | ||
AK012720.1 | - | 15 - 16 | 1860 | 61 | ||
AK014476.1 | - | 15 - 16 | 1851 | 61 | ||
AK031829.1 | - | 15 - 16 | 1886 | 61 | ||
AK032378.1 | - | 14 - 15 | 1970 | 61 | ||
AK162182.1 | - | 1643 | 61 | |||
BC058375.1 | - | 15 - 16 | 1748 | 6828178 | 61 | |
DQ278869.1 | - | 15 - 16 | 1883 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |