Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170486.1 | NP_001163957.1 | 2 - 3 | 206 | 91 | |
NM_026552.3 | NP_080828.1 | 3 - 4 | 325 | 91 | ||
GenBank mRNA | AJ278129.1 | - | 3 - 4 | 251 | 91 | |
AK017993.1 | - | 3 - 4 | 326 | 91 | ||
AK018069.1 | - | 3 - 4 | 263 | 91 | ||
AK039215.1 | - | 3 - 4 | 273 | 91 | ||
AK076248.1 | - | 3 - 4 | 273 | 91 | ||
AK077325.1 | - | 3 - 4 | 247 | 91 | ||
AK133626.1 | - | 3 - 4 | 273 | 91 | ||
AK136082.1 | - | 3 - 4 | 268 | 91 | ||
AK150690.1 | - | 3 - 4 | 259 | 91 | ||
AK153380.1 | - | 3 - 4 | 257 | 91 | ||
AK159410.1 | - | 2 - 3 | 154 | 91 | ||
AK166440.1 | - | 3 - 4 | 247 | 91 | ||
BC015280.1 | - | 2 - 3 | 237 | 91 | ||
BC055309.1 | - | 2 - 3 | 236 | 6529255 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |