Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026616.2 | NP_080892.1 | 1 - 2 | 211 | 74 | |
GenBank mRNA | AF346604.1 | - | 1 - 2 | 209 | 74 | |
AK011556.1 | - | 1 - 2 | 189 | 74 | ||
AK018776.1 | - | 1 - 2 | 200 | 74 | ||
BC024333.1 | - | 1 - 2 | 183 | 5040687 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |