Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027790.2 | NP_082066.2 | 1 - 2 | 64 | 113 | |
GenBank mRNA | AK017274.1 | - | 1 - 2 | 66 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Inventoried Probe spans exons |