Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027898.3 | NP_082174.3 | 3 - 4 | 388 | 78 | |
GenBank mRNA | AK004885.1 | - | 3 - 4 | 374 | 78 | |
AK083837.1 | - | 3 - 4 | 351 | 78 | ||
AK150032.1 | - | 3 - 4 | 327 | 78 | ||
AK150196.1 | - | 3 - 4 | 340 | 78 | ||
AK150837.1 | - | 3 - 4 | 354 | 78 | ||
AK151987.1 | - | 3 - 4 | 354 | 78 | ||
AK152294.1 | - | 3 - 4 | 354 | 78 | ||
AK154821.1 | - | 3 - 4 | 389 | 78 | ||
BC018554.1 | - | 2 - 3 | 147 | 2646082 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |