Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028186.4 | NP_082462.3 | 4 - 5 | 428 | 67 | |
GenBank mRNA | AF358136.1 | - | 4 - 5 | 378 | 67 | |
AK008814.1 | - | 4 - 5 | 410 | 67 | ||
AK081904.1 | - | 3 - 4 | 374 | 67 | ||
AK146667.1 | - | 4 - 5 | 410 | 67 | ||
AK158350.1 | - | 4 - 5 | 410 | 67 | ||
AK165116.1 | - | 3 - 4 | 374 | 67 | ||
AK199881.1 | - | 2 - 3 | 90 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |