Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028333.2 | NP_082609.2 | 1 - 2 | 214 | 95 | |
GenBank mRNA | AK012888.1 | - | 1 - 2 | 295 | 95 | |
AK142319.1 | - | 1 - 2 | 324 | 95 | ||
BC082563.1 | - | 1 - 2 | 214 | 30607749 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |