Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029091.2 | NP_083367.1 | 12 - 13 | 1554 | 72 | |
GenBank mRNA | AK004774.1 | - | 12 - 13 | 1555 | 72 | |
AK170793.1 | - | 12 - 13 | 1538 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |