Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171004.1 | NP_001164475.1 | 5 - 6 | 1476 | 70 | |
NM_001171005.1 | NP_001164476.1 | 4 - 5 | 672 | 70 | ||
NM_029239.3 | NP_083515.2 | 5 - 6 | 1476 | 70 | ||
GenBank mRNA | AK016167.1 | - | 5 - 6 | 631 | 70 | |
AK030058.1 | - | 4 - 5 | 669 | 70 | ||
AK050059.1 | - | 3 - 4 | 500 | 70 | ||
BC027320.1 | - | 4 - 5 | 875 | 70 | ||
BC037012.1 | - | 4 - 5 | 1120 | 5342428 | 70 | |
BC089164.1 | - | 5 - 6 | 1375 | 5400250 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |