Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029901.2 | NP_084177.2 | 2 - 3 | 331 | 89 | |
GenBank mRNA | AB178898.1 | - | 2 - 3 | 257 | 89 | |
AI882115.1 | - | 2 - 3 | 329 | 89 | ||
AK020439.1 | - | 2 - 3 | 331 | 89 | ||
AY742217.1 | - | 2 - 3 | 257 | 89 | ||
BC013531.1 | - | 2 - 3 | 307 | 4238756 | 89 | |
BC061057.1 | - | 2 - 3 | 327 | 30305353 | 89 | |
BC091761.1 | - | 2 - 3 | 307 | 5044038 | 89 | |
BY721648.1 | - | 2 - 3 | 331 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |