Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030127.3 | NP_084403.2 | 8 - 9 | 1649 | 76 | |
GenBank mRNA | AK035194.1 | - | 8 - 9 | 1652 | 76 | |
AY037300.1 | - | 8 - 9 | 1434 | 76 | ||
AY156509.1 | - | 8 - 9 | 1341 | 76 | ||
BC138587.1 | - | 8 - 9 | 1373 | 8861607 | 76 | |
BC138588.1 | - | 8 - 9 | 1373 | 8861608 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |