Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030558.2 | NP_085035.1 | 6 - 7 | 667 | 97 | |
GenBank mRNA | AF231122.1 | - | 6 - 7 | 619 | 97 | |
BC019975.1 | - | 6 - 7 | 667 | 4223351 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |