Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159603.1 | NP_001153075.1 | 16 - 17 | 2842 | 78 | |
NM_001159604.1 | NP_001153076.1 | 16 - 17 | 2842 | 78 | ||
NM_001159605.1 | NP_001153077.1 | 14 - 15 | 2554 | 78 | ||
NM_001159606.1 | NP_001153078.1 | 12 - 13 | 2116 | 78 | ||
NM_030722.2 | NP_109647.2 | 16 - 17 | 2845 | 78 | ||
GenBank mRNA | AF321909.1 | - | 16 - 17 | 2815 | 78 | |
AK122205.1 | - | 16 - 17 | 2825 | 78 | ||
AK141921.1 | - | 16 - 17 | 2842 | 78 | ||
AK147185.1 | - | 12 - 13 | 2088 | 78 | ||
AK154059.1 | - | 15 - 16 | 2440 | 78 | ||
AK161135.1 | - | 14 - 15 | 2548 | 78 | ||
AK162742.1 | - | 15 - 16 | 2538 | 78 | ||
AK163977.1 | - | 16 - 17 | 2833 | 78 | ||
BC048174.1 | - | 16 - 17 | 2795 | 6825203 | 78 | |
BC050747.1 | - | 16 - 17 | 2815 | 6593374 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |