Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1xr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030732.3 | NP_109657.2 | 9 - 10 | 1272 | 105 | |
GenBank mRNA | AF268195.1 | - | 9 - 10 | 1053 | 105 | |
AK029595.1 | - | 9 - 10 | 1071 | 105 | ||
AK033347.1 | - | 9 - 10 | 1142 | 105 | ||
AK036064.1 | - | 10 - 11 | 1383 | 105 | ||
AK136255.1 | - | 9 - 10 | 1094 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |