Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199293.1 | NP_001186222.1 | 15 - 16 | 2835 | 118 | |
NM_031386.2 | NP_113563.2 | 15 - 16 | 2816 | 118 | ||
GenBank mRNA | AK139808.1 | - | 2 - 3 | 572 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |