Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040085.2 | NP_001035174.1 | 16 - 17 | 3103 | 108 | |
NM_001040087.2 | NP_001035176.1 | 7 - 8 | 1310 | 108 | ||
NM_001040088.2 | NP_001035177.1 | 7 - 8 | 1238 | 108 | ||
NM_001289583.1 | NP_001276512.1 | 17 - 18 | 3151 | 108 | ||
NM_001289584.1 | NP_001276513.1 | 17 - 18 | 3070 | 108 | ||
NM_001289585.1 | NP_001276514.1 | 8 - 9 | 1403 | 108 | ||
NM_001289586.1 | NP_001276515.1 | 8 - 9 | 1358 | 108 | ||
NM_031394.3 | NP_113571.2 | 16 - 17 | 3031 | 108 | ||
GenBank mRNA | AB050742.1 | - | 7 - 8 | 849 | 108 | |
AB057754.1 | - | 16 - 17 | 2490 | 108 | ||
AB057755.1 | - | 10 - 11 | 3000 | 108 | ||
AB057756.1 | - | 8 - 9 | 1325 | 108 | ||
AB057757.1 | - | 3 - 4 | 1316 | 108 | ||
AB057760.1 | - | 16 - 17 | 2409 | 108 | ||
AB057761.1 | - | 15 - 16 | 2442 | 108 | ||
AB057762.1 | - | 15 - 16 | 2370 | 108 | ||
AB057763.1 | - | 4 - 5 | 1219 | 108 | ||
AK027924.1 | - | 7 - 8 | 1221 | 108 | ||
AK051653.1 | - | 8 - 9 | 1404 | 108 | ||
AK173212.1 | - | 7 - 8 | 768 | 108 | ||
BC060665.1 | - | 8 - 9 | 2853 | 108 | ||
BC138714.1 | - | 15 - 16 | 2341 | 8861736 | 108 | |
BC145398.1 | - | 16 - 17 | 2461 | 9053956 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |