Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031395.2 | NP_113572.1 | 12 - 13 | 1682 | 69 | |
NM_183368.2 | NP_899224.2 | 6 - 7 | 878 | 69 | ||
NM_183370.2 | NP_899226.2 | 10 - 11 | 1096 | 69 | ||
GenBank mRNA | AB050743.1 | - | 914 | 69 | ||
AB057758.1 | - | 13 - 14 | 1682 | 69 | ||
AB057764.1 | - | 14 - 15 | 1826 | 69 | ||
AB057765.1 | - | 12 - 13 | 1603 | 69 | ||
AB057766.1 | - | 1024 | 69 | |||
AK131174.1 | - | 11 - 12 | 6437 | 69 | ||
BB031741.2 | - | 1 - 2 | 48 | 69 | ||
BC022608.1 | - | 11 - 12 | 1305 | 4166796 | 69 | |
BC106102.1 | - | 10 - 11 | 1096 | 3980415 | 69 | |
BC118936.2 | - | 13 - 14 | 1507 | 40091060 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |