Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031842.2 | NP_114030.2 | 4 - 5 | 565 | 83 | |
GenBank mRNA | BC026783.1 | - | 4 - 5 | 477 | 4481097 | 83 |
BC059921.1 | - | 4 - 5 | 535 | 83 | ||
BC079839.1 | - | 4 - 5 | 511 | 6810462 | 83 | |
CV562378.1 | - | 4 - 5 | 565 | 83 | ||
M25773.1 | - | 4 - 5 | 521 | 83 | ||
U66620.1 | - | 4 - 5 | 521 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |