Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033074.3 | NP_149065.2 | 5 - 6 | 693 | 83 | |
GenBank mRNA | AK011146.2 | - | 5 - 6 | 694 | 83 | |
AK031064.1 | - | 5 - 6 | 657 | 83 | ||
AK152945.1 | - | 5 - 6 | 654 | 83 | ||
AK153314.1 | - | 5 - 6 | 665 | 83 | ||
AK166693.1 | - | 5 - 6 | 660 | 83 | ||
AK167597.1 | - | 5 - 6 | 702 | 83 | ||
AK168969.1 | - | 5 - 6 | 653 | 83 | ||
BC055371.1 | - | 5 - 6 | 679 | 30060749 | 83 | |
BC060176.1 | - | 632 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |