Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033079.2 | NP_149070.2 | 5 - 6 | 851 | 75 | |
GenBank mRNA | AF084364.1 | - | 4 - 5 | 820 | 75 | |
AK131954.1 | - | 5 - 6 | 851 | 75 | ||
BC119803.1 | - | 4 - 5 | 778 | 40107692 | 75 | |
BC122521.1 | - | 4 - 5 | 783 | 40107688 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |