Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033079.2 | NP_149070.2 | 7 - 8 | 1167 | 65 | |
GenBank mRNA | AF084364.1 | - | 6 - 7 | 1136 | 65 | |
AK131954.1 | - | 7 - 8 | 1167 | 65 | ||
BC119803.1 | - | 6 - 7 | 1094 | 40107692 | 65 | |
BC122521.1 | - | 6 - 7 | 1099 | 40107688 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |