Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033268.4 | NP_150371.4 | 8 - 9 | 1014 | 99 | |
GenBank mRNA | AF248643.1 | - | 8 - 9 | 782 | 99 | |
AY036877.1 | - | 9 - 10 | 958 | 99 | ||
BC089579.1 | - | 8 - 9 | 1014 | 6757718 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |