Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arvcf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272028.1 | NP_001258957.1 | 14 - 15 | 2834 | 57 | |
NM_001272029.1 | NP_001258958.1 | 14 - 15 | 2967 | 57 | ||
NM_001272030.1 | NP_001258959.1 | 13 - 14 | 2816 | 57 | ||
NM_001272031.1 | NP_001258960.1 | 13 - 14 | 2577 | 57 | ||
NM_001272032.1 | NP_001258961.1 | 12 - 13 | 2559 | 57 | ||
NM_033474.3 | NP_258435.2 | 15 - 16 | 2985 | 57 | ||
GenBank mRNA | AF286212.1 | - | 14 - 15 | 2834 | 57 | |
AF286213.1 | - | 13 - 14 | 2816 | 57 | ||
AF286214.1 | - | 13 - 14 | 2486 | 57 | ||
AF286215.1 | - | 12 - 13 | 2468 | 57 | ||
AJ243418.1 | - | 13 - 14 | 2601 | 57 | ||
AK031855.1 | - | 8 - 9 | 942 | 57 | ||
AK044982.1 | - | 12 - 13 | 2507 | 57 | ||
AK051627.1 | - | 13 - 14 | 2533 | 57 | ||
AK084886.1 | - | 14 - 15 | 2955 | 57 | ||
BC051655.1 | - | 9 - 10 | 982 | 57 | ||
BC052393.1 | - | 14 - 15 | 2886 | 57 | ||
BC056980.1 | - | 15 - 16 | 2893 | 6821090 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |