Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arvcf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272028.1 | NP_001258957.1 | 15 - 16 | 2889 | 73 | |
NM_001272029.1 | NP_001258958.1 | 15 - 16 | 3022 | 73 | ||
NM_001272030.1 | NP_001258959.1 | 14 - 15 | 2871 | 73 | ||
NM_001272031.1 | NP_001258960.1 | 14 - 15 | 2632 | 73 | ||
NM_001272032.1 | NP_001258961.1 | 13 - 14 | 2614 | 73 | ||
NM_033474.3 | NP_258435.2 | 16 - 17 | 3040 | 73 | ||
GenBank mRNA | AF286212.1 | - | 15 - 16 | 2889 | 73 | |
AF286213.1 | - | 14 - 15 | 2871 | 73 | ||
AF286214.1 | - | 14 - 15 | 2541 | 73 | ||
AF286215.1 | - | 13 - 14 | 2523 | 73 | ||
AJ243418.1 | - | 14 - 15 | 2656 | 73 | ||
AK031855.1 | - | 9 - 10 | 997 | 73 | ||
AK044982.1 | - | 13 - 14 | 2562 | 73 | ||
AK051627.1 | - | 14 - 15 | 2588 | 73 | ||
AK084886.1 | - | 15 - 16 | 3010 | 73 | ||
BC051655.1 | - | 10 - 11 | 1037 | 73 | ||
BC052393.1 | - | 15 - 16 | 2941 | 73 | ||
BC056980.1 | - | 16 - 17 | 2948 | 6821090 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |